This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRSS30 SIRNA Assays ›
Gene Symbol:
Prss30
Gene Name:
protease, serine, 30
Gene Aliases:
Tmprss8, tmsp-1
Chromosome Location:
Chr.10: 13348289 - 13355837 on Build Rnor_6.0
UniGene ID:
Rn.102783
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 196935

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prss30
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199371.1 NP_955403.1 6 1102
XM_006245922.2 XP_006245984.1 7 1293
XM_006245923.2 XP_006245985.1 7 1301
XM_008767553.1 XP_008765775.1 7 1321
GenBank mRNA AB073023.1 - 6 1102

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
sodium ion transport
serine-type endopeptidase activity
sodium channel regulator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0