This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other USP48 SIRNA Assays ›
Gene Symbol:
Usp48
Gene Name:
ubiquitin specific peptidase 48
Gene Aliases:
Usp31, synUSP
Chromosome Location:
Chr.5: 155935105 - 156002722 on Build Rnor_6.0
UniGene ID:
Rn.100022
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 196775

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Usp48
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198785.1 NP_942080.1 2 333
XM_006239234.2 XP_006239296.1 3 367
XM_006239235.2 XP_006239297.1 3 367
XM_006239236.2 XP_006239298.1 3 367
XM_006239237.2 XP_006239299.1 3 367
GenBank mRNA AB073880.1 - 2 333

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0