This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IGF2BP1 SIRNA Assays ›
Gene Symbol:
Igf2bp1
Gene Name:
insulin-like growth factor 2 mRNA binding protein 1
Gene Aliases:
Imp1, ZBP1
Chromosome Location:
Chr.10: 83774178 - 83823317 on Build Rnor_6.0
UniGene ID:
Rn.77572
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 196464

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Igf2bp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175594.2 NP_783184.1 5, 6 633
XM_006247193.2 XP_006247255.1 5, 6 954
GenBank mRNA AF541940.1 - 5, 6 633
BC161831.1 - 5, 6 665 9023972

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

RNA localization
regulation of mRNA stability involved in response to stress
negative regulation of translation
pallium cell proliferation in forebrain
mRNA transport
CRD-mediated mRNA stabilization
neuronal stem cell population maintenance
nucleotide binding
mRNA binding
mRNA 3'-UTR binding
poly(A) RNA binding
translation regulator activity
mRNA 5'-UTR binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0