This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PEX14 SIRNA Assays ›
Gene Symbol:
Pex14
Gene Name:
peroxisomal biogenesis factor 14
Gene Aliases:
-
Chromosome Location:
Chr.5: 165782895 - 165918445 on Build Rnor_6.0
UniGene ID:
Rn.7844
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 196406

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pex14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172063.1 NP_742060.1 4 303
GenBank mRNA AB017544.1 - 4 303
BC081708.1 - 4 292 7191447

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein complex assembly
peroxisome organization
protein import into peroxisome matrix
protein import into peroxisome matrix, docking
protein import into peroxisome matrix, translocation
negative regulation of protein binding
microtubule anchoring
peroxisome transport along microtubule
negative regulation of sequence-specific DNA binding transcription factor activity
protein import into peroxisome matrix, substrate release
negative regulation of transcription, DNA-templated
protein homooligomerization
negative regulation of protein homotetramerization
transcription corepressor activity
receptor binding
microtubule binding
protein N-terminus binding
beta-tubulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0