This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OPTN SIRNA Assays ›
Gene Symbol:
Optn
Gene Name:
optineurin
Gene Aliases:
Fip2
Chromosome Location:
Chr.17: 77167701 - 77218374 on Build Rnor_6.0
UniGene ID:
Rn.63435
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 196320

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Optn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145081.3 NP_659549.2 1 127
XM_006254262.2 XP_006254324.1 2 285
XM_006254263.2 XP_006254325.1 2 284
XM_006254264.2 XP_006254326.1 2 284
XM_008771852.1 XP_008770074.1 2 285
XM_008771853.1 XP_008770075.1 2 284
GenBank mRNA AB069907.3 - 1 127
AB222074.1 - 1 81
BC086976.1 - 2 213 7111026
FQ223875.1 - 2 284

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting to Golgi
negative regulation of receptor recycling
autophagy
Golgi organization
positive regulation of cell proliferation
macroautophagy
cellular protein localization
wound healing
Golgi to plasma membrane protein transport
regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of neuron apoptotic process
cytokine secretion
defense response to Gram-negative bacterium
protein homooligomerization
Golgi ribbon formation
TFIIIA-class transcription factor binding
protein C-terminus binding
Rab GTPase binding
polyubiquitin binding
identical protein binding
ubiquitin binding
K63-linked polyubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0