This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRDM4 SIRNA Assays ›
Gene Symbol:
Prdm4
Gene Name:
PR domain containing 4
Gene Aliases:
SC-1
Chromosome Location:
Chr.7: 23959929 - 23986535 on Build Rnor_6.0
UniGene ID:
Rn.162921
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 196134

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prdm4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133312.1 NP_579846.1 3, 4 1108
XM_006241160.2 XP_006241222.1 5, 6 1570
XM_006241161.1 XP_006241223.1 5, 6 1569
XM_008765213.1 XP_008763435.1 5, 6 1575
GenBank mRNA AF176023.1 - 3, 4 1108

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_012406, AL133083, AF144757, BC035581, AK056378.
This siRNA also targets the following transcripts in Mouse: NM_181650, AK028414, AK029718, BC042516, BC025642, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of cell growth
cellular response to nutrient
methylation
cellular response to growth factor stimulus
DNA binding
death receptor binding
methyltransferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0