This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DECR1 SIRNA Assays ›
Gene Symbol:
Decr1
Gene Name:
2,4-dienoyl CoA reductase 1, mitochondrial
Gene Aliases:
-
Chromosome Location:
Chr.5: 29573893 - 29601731 on Build Rnor_6.0
UniGene ID:
Rn.2854
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 195946

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Decr1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057197.2 NP_476545.2 2 139
GenBank mRNA BC059120.1 - 2 127 6920489
CO385941.1 - 2 124
D00569.1 - 2 112
FQ209689.1 - 2 128
FQ210607.1 - 2 120
FQ210721.1 - 2 122
FQ210830.1 - 2 109
FQ212507.1 - 2 117
FQ218387.1 - 2 117
FQ219629.1 - 2 120
FQ219666.1 - 2 117
FQ220227.1 - 2 120

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
protein homotetramerization
2,4-dienoyl-CoA reductase (NADPH) activity
NADPH binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0