This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SERPINB5 SIRNA Assays ›
Gene Symbol:
Serpinb5
Gene Name:
serpin peptidase inhibitor, clade B (ovalbumin), member 5
Gene Aliases:
Maspin, PI-5, Pi5
Chromosome Location:
Chr.13: 26903052 - 26923253 on Build Rnor_6.0
UniGene ID:
Rn.25752
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 195898

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Serpinb5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057108.2 NP_476449.1 2, 3 283
XM_008769405.1 XP_008767627.1 1, 2 441

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009257, U54705, BC005434, AK136532, AK145220.

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelium
negative regulation of endopeptidase activity
extracellular matrix organization
regulation of epithelial cell proliferation
prostate gland morphogenesis
serine-type endopeptidase inhibitor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0