This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LANCL1 SIRNA Assays ›
Gene Symbol:
Lancl1
Gene Name:
LanC lantibiotic synthetase component C-like 1 (bacterial)
Gene Aliases:
GPR69A, p40
Chromosome Location:
Chr.9: 74018192 - 74048098 on Build Rnor_6.0
UniGene ID:
Rn.38972
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 195706

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lancl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053723.1 NP_446175.1 9 1291
GenBank mRNA AJ131111.1 - 9 1291
BC085811.1 - 9 1269 7115423
FQ211687.1 - 4 396

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
metabolic process
catalytic activity
zinc ion binding
SH3 domain binding
glutathione binding
low-density lipoprotein particle receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0