This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MADD SIRNA Assays ›
Gene Symbol:
Madd
Gene Name:
MAP-kinase activating death domain
Gene Aliases:
-
Chromosome Location:
Chr.3: 79960301 - 80003023 on Build Rnor_6.0
UniGene ID:
Rn.90117
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 195619

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Madd
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053585.1 NP_446037.1 4 1030
GenBank mRNA U72995.1 - 4 1030

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001177719, NM_145527, NM_001177720, AK036347, AY263984, and others.

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
regulation of Rab protein signal transduction
regulation of apoptotic process
regulation of cell cycle
negative regulation of growth hormone secretion
execution phase of apoptosis
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of pancreatic amylase secretion
negative regulation of apoptotic signaling pathway
regulation of extrinsic apoptotic signaling pathway
protein binding
Rab guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0