This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDC5L SIRNA Assays ›
Gene Symbol:
Cdc5l
Gene Name:
cell division cycle 5-like
Gene Aliases:
-
Chromosome Location:
Chr.9: 17949764 - 17988514 on Build Rnor_6.0
UniGene ID:
Rn.54977
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 195574

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdc5l
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053527.1 NP_445979.1 10 1439
GenBank mRNA AF000578.2 - 10 1439
BC087007.1 - 11 1486
BC161839.1 - 10 1403 9101355

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA splicing
signal transduction involved in DNA damage checkpoint
DNA binding
chromatin binding
protein binding
protein domain specific binding
poly(A) RNA binding
WD40-repeat domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm