This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HAO2 SIRNA Assays ›
Gene Symbol:
Hao2
Gene Name:
hydroxyacid oxidase 2 (long chain)
Gene Aliases:
Hao3
Chromosome Location:
Chr.2: 200785492 - 200808849 on Build Rnor_6.0
UniGene ID:
Rn.198611
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 195399

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hao2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032082.2 NP_114471.1 9 1462
XM_006233022.2 XP_006233084.1 10 1627
XM_006233024.2 XP_006233086.1 10 1618
XM_006233025.2 XP_006233087.1 9 1486
GenBank mRNA BC078781.1 - 8 1378 7132928
FQ210841.1 - 1 217
X67156.1 - 9 1463

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mandelate metabolic process
fatty acid oxidation
protein homooligomerization
(S)-2-hydroxy-acid oxidase activity
receptor binding
FMN binding
very-long-chain-(S)-2-hydroxy-acid oxidase activity
long-chain-(S)-2-hydroxy-long-chain-acid oxidase activity
medium-chain-(S)-2-hydroxy-acid oxidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0