This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD164 SIRNA Assays ›
Gene Symbol:
Cd164
Gene Name:
CD164 molecule, sialomucin
Gene Aliases:
MGC-24v
Chromosome Location:
Chr.20: 46250418 - 46261994 on Build Rnor_6.0
UniGene ID:
Rn.3733
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 195321

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd164
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031812.1 NP_114000.1 3 285
GenBank mRNA AJ238574.1 - 3 285
BC062064.1 - 3 288 5599253

Back To Top

More Information


Gene Ontology Categories:

Function(s)

cell adhesion
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
muscle organ development

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0