This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STK3 SIRNA Assays ›
Gene Symbol:
Stk3
Gene Name:
serine/threonine kinase 3
Gene Aliases:
MST2
Chromosome Location:
Chr.7: 73617926 - 73883885 on Build Rnor_6.0
UniGene ID:
Rn.89363
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 195221

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stk3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031735.1 NP_113923.1 2, 3 190
XM_008765458.1 XP_008763680.1 2, 3 368
XM_008765459.1 XP_008763681.1 2, 3 368
XM_008765461.1 XP_008763683.1 2, 3 368
XM_008765462.1 XP_008763684.1 2, 3 368
GenBank mRNA AJ001529.1 - 2, 3 190
BC161844.1 - 2, 3 273 9027884

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006281, NM_001256312, NM_001256313, AK312660, AK075229, and others.
This siRNA also targets the following transcripts in Mouse: NM_019635, U28726, AF271361, BC049123, AY058922.

Gene Ontology Categories:

Function(s) Process(es)

neural tube formation
endocardium development
protein phosphorylation
apoptotic process
central nervous system development
negative regulation of cell proliferation
signal transduction by protein phosphorylation
positive regulation of protein binding
hippo signaling
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of fat cell differentiation
positive regulation of JNK cascade
negative regulation of organ growth
protein stabilization
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of protein kinase B signaling
primitive hemopoiesis
cell differentiation involved in embryonic placenta development
regulation of cell differentiation involved in embryonic placenta development
negative regulation of canonical Wnt signaling pathway
hepatocyte apoptotic process
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
magnesium ion binding
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
ATP binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0