This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MYH10 SIRNA Assays ›
Gene Symbol:
Myh10
Gene Name:
myosin, heavy chain 10, non-muscle
Gene Aliases:
MCH-B, NMMHC-B, SMemb
Chromosome Location:
Chr.10: 55274914 - 55406738 on Build Rnor_6.0
UniGene ID:
Rn.98166
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 195047

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Myh10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031520.1 NP_113708.1 6 857
XM_006246821.2 XP_006246883.1 6 904
XM_008767880.1 XP_008766102.1 6 1014
XM_008767881.1 XP_008766103.1 6 730
GenBank mRNA AF139055.1 - 6 857

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001256012, NM_005964, NM_001256095, U15618, BC117691, and others.
This siRNA also targets the following transcripts in Mouse: NM_175260, AK045066, AK147567, AK163604, AK133946, and others.

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
in utero embryonic development
neuron migration
plasma membrane repair
exocytosis
substrate-dependent cell migration, cell extension
nuclear migration
cell adhesion
axonogenesis
axon guidance
brain development
adult heart development
metabolic process
cell proliferation
regulation of cell shape
fourth ventricle development
lateral ventricle development
third ventricle development
cerebellar Purkinje cell layer development
actin cytoskeleton organization
actin filament-based movement
myofibril assembly
actomyosin structure organization
neuron projection development
neuromuscular process controlling balance
cardiac myofibril assembly
ventricular cardiac muscle cell development
retina development in camera-type eye
microfilament motor activity
motor activity
protein binding
calmodulin binding
ATP binding
actin-dependent ATPase activity
ADP binding
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0