This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DBN1 SIRNA Assays ›
Gene Symbol:
Dbn1
Gene Name:
drebrin 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 9679526 - 9693878 on Build Rnor_6.0
UniGene ID:
Rn.11247
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 194931

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dbn1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031024.1 NP_112286.1 13 1804
XM_006253655.2 XP_006253717.1 13 1890
XM_006253656.2 XP_006253718.1 12 1751
XM_006253657.2 XP_006253719.1 12 1750
XM_008771507.1 XP_008769729.1 12 2356
GenBank mRNA AB015042.1 - 12 1613
AB514558.1 - 12 1613
BC139847.1 - 12 1668 8368018
FQ217148.1 - 12 1695
X59267.1 - 13 1804

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
nervous system development
cell communication by chemical coupling
cell communication by electrical coupling
maintenance of protein location in cell
generation of neurons
neural precursor cell proliferation
actin binding
protein binding
profilin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0