This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other INPPL1 SIRNA Assays ›
Gene Symbol:
Inppl1
Gene Name:
inositol polyphosphate phosphatase-like 1
Gene Aliases:
Ship2
Chromosome Location:
Chr.1: 166898177 - 166912524 on Build Rnor_6.0
UniGene ID:
Rn.42902
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 194746

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Inppl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270843.1 NP_001257772.1 12 1557
NM_022944.2 NP_075233.1 13 1613
GenBank mRNA AB011439.1 - 12 1557
AB025794.1 - 13 1611

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

endochondral ossification
immune system process
glucose metabolic process
phosphatidylinositol biosynthetic process
endocytosis
actin filament organization
cell adhesion
brain development
negative regulation of DNA replication
negative regulation of cell proliferation
post-embryonic development
negative regulation of gene expression
negative regulation of platelet-derived growth factor receptor signaling pathway
negative regulation of neuron projection development
response to insulin
inositol trisphosphate metabolic process
response to drug
negative regulation of MAP kinase activity
negative regulation of insulin-like growth factor receptor signaling pathway
cellular lipid metabolic process
negative regulation of insulin receptor signaling pathway
phosphatidylinositol dephosphorylation
ruffle assembly
actin binding
inositol-polyphosphate 5-phosphatase activity
protein binding
phosphatidylinositol-3,4,5-trisphosphate binding
SH3 domain binding
SH2 domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0