This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SHOC2 SIRNA Assays ›
Gene Symbol:
SHOC2
Gene Name:
SHOC2, leucine rich repeat scaffold protein
Gene Aliases:
SIAA0862, SOC2, SUR8
Chromosome Location:
Chr.10: 110919370 - 111013667 on Build GRCh38
UniGene ID:
Hs.104315
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 19469

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SHOC2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001269039.2 NP_001255968.1 2 435
NM_001324336.1 NP_001311265.1 2 392
NM_001324337.1 NP_001311266.1 3 620
NM_007373.3 NP_031399.2 2 435
XM_017016702.1 XP_016872191.1 2 340
XM_017016703.1 XP_016872192.1 2 6157
XM_017016704.1 XP_016872193.1 2 339
GenBank mRNA AB020669.1 - 2 363
AF054828.1 - 2 357
AF068920.1 - 1 86
AK292833.1 - 3 561
BC044752.1 - 2 371
BC050445.1 - 1 290 6067669
BX649055.1 - 2 361

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001013155, BC079032.

Gene Ontology Categories:

Function(s) Process(es)

Ras protein signal transduction
fibroblast growth factor receptor signaling pathway
positive regulation of Ras protein signal transduction
regulation of catalytic activity
protein binding
protein phosphatase 1 binding
protein phosphatase regulator activity
protein phosphatase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0