This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other METAP2 SIRNA Assays ›
Gene Symbol:
Metap2 RGD1560341
Gene Name:
methionyl aminopeptidase 2
similar to Methionine aminopeptidase 2 (MetAP 2)
Gene Aliases:
Amp2, Mnpep, p67
-
Chromosome Location:
Chr.7: 34762241 - 34789456 on Build Rnor_6.0
Chromosome Location:
Chr.18: 51218243 - 51220149 on Build Rnor_6.0
UniGene ID:
Rn.1984
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 194633

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Metap2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022539.1 NP_071984.1 5, 6 855
GenBank mRNA BC070892.1 - 5, 6 770 7099147
BC091150.1 - 5, 6 869 7122560
L10652.1 - 5, 6 855
Gene Symbol: RGD1560341
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_574154.6 XP_574154.2 1 885

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
protein processing
peptidyl-methionine modification
N-terminal protein amino acid modification
protein initiator methionine removal
aminopeptidase activity
metalloexopeptidase activity
poly(A) RNA binding
metal ion binding
metalloaminopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0