This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BCAS1 SIRNA Assays ›
Gene Symbol:
Bcas1
Gene Name:
breast carcinoma amplified sequence 1
Gene Aliases:
Band83, Pmes-2
Chromosome Location:
Chr.3: 167957994 - 168033457 on Build Rnor_6.0
UniGene ID:
Rn.154429
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 193951

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bcas1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762466.1 XP_008760688.1 6 1033
GenBank mRNA AB029897.1 - 4 541
AB180269.1 - 4 541
AB180270.1 - 4 541
AB180271.1 - 4 541
BC094306.1 - 4 529
CO405877.1 - 2 127

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029815, AK008957, AY219233.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0