This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LMBRD1 SIRNA Assays ›
Gene Symbol:
Lmbrd1
Gene Name:
LMBR1 domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 30939555 - 31038381 on Build Rnor_6.0
UniGene ID:
Rn.234342
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 193826

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lmbrd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139189.2 NP_631928.2 2 205
GenBank mRNA AY093598.1 - 2 117
BC078723.1 - 2 205 7123105

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

insulin receptor internalization
regulation of MAPK cascade
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
negative regulation of protein kinase B signaling
insulin receptor binding
cobalamin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0