This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DMGDH SIRNA Assays ›
Gene Symbol:
Dmgdh
Gene Name:
dimethylglycine dehydrogenase
Gene Aliases:
Me2GlyDH
Chromosome Location:
Chr.2: 23289376 - 23370360 on Build Rnor_6.0
UniGene ID:
Rn.3646
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 193793

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dmgdh
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139102.2 NP_620802.2 2 167
XM_008760629.1 XP_008758851.1 2 182
XM_008760630.1 XP_008758852.1 2 182
XM_008760631.1 XP_008758853.1 2 182
XM_008760632.1 XP_008758854.1 2 182
GenBank mRNA BC085697.1 - 2 167 7108858
X55995.1 - 2 135

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028772, AK004755, BC089599.

Gene Ontology Categories:

Function(s) Process(es)

glycine metabolic process
amino-acid betaine catabolic process
choline metabolic process
tetrahydrofolate interconversion
choline catabolic process
oxidation-reduction process
folic acid binding
poly(A) RNA binding
dimethylglycine dehydrogenase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0