This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DNAJC21 SIRNA Assays ›
Gene Symbol:
Dnajc21
Gene Name:
DnaJ (Hsp40) homolog, subfamily C, member 21
Gene Aliases:
Dnaja5
Chromosome Location:
Chr.2: 60419446 - 60446666 on Build Rnor_6.0
UniGene ID:
Rn.53644
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 193630

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dnajc21
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138856.2 NP_620211.2 2, 3 219
XM_008760746.1 XP_008758968.1 6, 7 1095
GenBank mRNA AF492384.1 - 1, 2 142
BC086587.1 - 2, 3 219
BC158554.1 - 2, 3 438 7380100

Back To Top

More Information


Gene Ontology Categories:

Process(es)

zinc ion binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0