This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100909664 SIRNA Assays ›
Gene Symbol:
LOC100909664 Cep170
Gene Name:
centrosomal protein of 170 kDa-like
centrosomal protein 170
Gene Aliases:
-
KAB1, Kab, Rhk1
Chromosome Location:
Chr.13: 43806604 - 43845827 on Build Rnor_6.0
Chromosome Location:
Chr.13: 94807090 - 94887448 on Build Rnor_6.0
UniGene ID:
-
Rn.6424
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 193536

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100909664
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751234.3 XP_003751282.2 9 2652
XM_006249680.2 XP_006249742.1 8 2839
XM_006249681.2 XP_006249743.1 8 2839
Gene Symbol: Cep170
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769812.1 XP_008768034.1 9 1757
GenBank mRNA AB038495.1 - 5 1614

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0