This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BZW2 SIRNA Assays ›
Gene Symbol:
Bzw2
Gene Name:
basic leucine zipper and W2 domains 2
Gene Aliases:
Hfb2
Chromosome Location:
Chr.6: 55586754 - 55647650 on Build Rnor_6.0
UniGene ID:
Rn.13196
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 193501

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bzw2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134402.3 NP_599229.1 11 1331
GenBank mRNA AF031483.1 - 11 1359
BC063149.1 - 11 1335 6890706
FQ225034.1 - 11 1307
FQ228189.1 - 11 1308

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025840, AK160544, AK178264, AK003229, BC013060, and others.

Gene Ontology Categories:

Function(s)

nervous system development
cell differentiation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0