This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CLCC1 SIRNA Assays ›
Gene Symbol:
Clcc1
Gene Name:
chloride channel CLIC-like 1
Gene Aliases:
Mclc
Chromosome Location:
Chr.2: 211450437 - 211479885 on Build Rnor_6.0
UniGene ID:
Rn.26586
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 193295

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Clcc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133414.1 NP_596905.1 9 985
XM_006233127.1 XP_006233189.1 9 1024
XM_006233128.2 XP_006233190.1 9 1083
XM_008761365.1 XP_008759587.1 9 1083
XM_008761366.1 XP_008759588.1 8 922
GenBank mRNA AB052919.1 - 7 841
AB052920.1 - 7 841
AB052921.1 - 8 943
AB052922.1 - 9 985
AB052923.1 - 9 1005
AF117330.1 - 9 973
BC081736.1 - 9 1061 7128078

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chloride transport
chloride transmembrane transport
chloride channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0