This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC36A1 SIRNA Assays ›
Gene Symbol:
Slc36a1
Gene Name:
solute carrier family 36 (proton/amino acid symporter), member 1
Gene Aliases:
Lyaat1
Chromosome Location:
Chr.10: 40538013 - 40573304 on Build Rnor_6.0
UniGene ID:
Rn.76305
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 193219

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc36a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130415.1 NP_569099.1 7 848
XM_008767657.1 XP_008765879.1 6 2947
XM_008767659.1 XP_008765881.1 8 1530
GenBank mRNA AF361239.1 - 7 848

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

neutral amino acid transport
L-alanine transport
glycine transport
proline transport
proton transport
proline transmembrane transport
L-alpha-amino acid transmembrane transport
hydrogen ion transmembrane transport
hydrogen:amino acid symporter activity
L-tyrosine transmembrane transporter activity
hydrogen ion transmembrane transporter activity
neutral amino acid transmembrane transporter activity
L-alanine transmembrane transporter activity
glycine transmembrane transporter activity
L-proline transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0