This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DNASE1L3 SIRNA Assays ›
Gene Symbol:
Dnase1l3
Gene Name:
deoxyribonuclease 1-like 3
Gene Aliases:
-
Chromosome Location:
Chr.15: 18710492 - 18736476 on Build Rnor_6.0
UniGene ID:
Rn.29996
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 193030

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dnase1l3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053907.1 NP_446359.1 3 470
GenBank mRNA AF039852.1 - 3 420
BC088122.1 - 3 468 7325326
FQ226037.1 - 3 297
FQ227612.1 - 3 297
FQ230283.1 - 3 297
FQ230990.1 - 3 297
FQ231646.1 - 3 296
FQ231744.1 - 3 296
FQ231870.1 - 3 300
FQ231939.1 - 3 297
FQ233228.1 - 3 353
FQ234709.1 - 3 353
U75689.1 - 3 470

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

apoptotic DNA fragmentation
programmed cell death involved in cell development
DNA binding
endonuclease activity
endodeoxyribonuclease activity
endodeoxyribonuclease activity, producing 5'-phosphomonoesters

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0