This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other F2RL1 SIRNA Assays ›
Gene Symbol:
F2rl1
Gene Name:
coagulation factor II (thrombin) receptor-like 1
Gene Aliases:
Par-2, Par2
Chromosome Location:
Chr.2: 25222324 - 25235275 on Build Rnor_6.0
UniGene ID:
Rn.10543
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 193013

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: F2rl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053897.2 NP_446349.1 2 1267
GenBank mRNA BC099765.1 - 2 1267 7930573
U61373.1 - 2 1180

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007974, Z48043, AX785009, AK144986, BC025432, and others.

Gene Ontology Categories:

Function(s) Process(es)

T cell activation involved in immune response
positive regulation of leukocyte chemotaxis
positive regulation of cytokine secretion involved in immune response
positive regulation of glomerular filtration
inflammatory response
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
blood coagulation
protein secretion
negative regulation of tumor necrosis factor-mediated signaling pathway
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of blood coagulation
positive regulation of cell migration
positive regulation of actin filament depolymerization
positive regulation of pseudopodium assembly
positive regulation of superoxide anion generation
positive regulation of toll-like receptor 2 signaling pathway
negative regulation of toll-like receptor 3 signaling pathway
positive regulation of toll-like receptor 3 signaling pathway
positive regulation of toll-like receptor 4 signaling pathway
positive regulation of Rho protein signal transduction
chemokine (C-C motif) ligand 2 secretion
neutrophil activation
regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of eosinophil degranulation
innate immune response
positive regulation of vasodilation
positive regulation of transcription from RNA polymerase II promoter
regulation of JNK cascade
negative regulation of JNK cascade
positive regulation of JNK cascade
interleukin-1 beta secretion
leukocyte migration
positive regulation of chemotaxis
positive regulation of positive chemotaxis
defense response to virus
positive regulation of phagocytosis, engulfment
establishment of endothelial barrier
positive regulation of ERK1 and ERK2 cascade
thrombin receptor signaling pathway
leukocyte proliferation
positive regulation of neutrophil mediated killing of gram-negative bacterium
interleukin-10 secretion
interferon-gamma secretion
chemokine secretion
negative regulation of chemokine secretion
mature conventional dendritic cell differentiation
positive regulation of renin secretion into blood stream
positive regulation of interleukin-8 secretion
positive regulation of interleukin-6 secretion
G-protein alpha-subunit binding
G-protein coupled receptor activity
protein binding
thrombin receptor activity
G-protein beta-subunit binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0