This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PLCB2 SIRNA Assays ›
Gene Symbol:
Plcb2
Gene Name:
phospholipase C, beta 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 110497760 - 110517261 on Build Rnor_6.0
UniGene ID:
Rn.30033
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 192753

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Plcb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053478.1 NP_445930.1 5 539
GenBank mRNA AJ011035.1 - 5 539
DQ120505.1 - 5 465
DQ120506.1 - 5 465

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

detection of chemical stimulus involved in sensory perception of bitter taste
metabolic process
phospholipid catabolic process
intracellular signal transduction
sensory perception of bitter taste
phosphatidylinositol phospholipase C activity
phospholipase C activity
signal transducer activity
calcium ion binding
phospholipid binding
G-protein beta/gamma-subunit complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0