This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STX3 SIRNA Assays ›
Gene Symbol:
Stx3
Gene Name:
syntaxin 3
Gene Aliases:
Stx3a
Chromosome Location:
Chr.1: 228137230 - 228194989 on Build Rnor_6.0
UniGene ID:
Rn.10951
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 192514

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stx3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031124.1 NP_112386.1 3 210
XM_006231097.2 XP_006231159.1 3 294
XM_008760262.1 XP_008758484.1 3 294
XM_008760263.1 XP_008758485.1 3 294
XM_008760264.1 XP_008758486.1 3 294
XM_008760265.1 XP_008758487.1 3 294
XM_008760266.1 XP_008758488.1 3 294
XM_008760267.1 XP_008758489.1 3 294
GenBank mRNA BC168246.1 - 3 164 5600098
L20820.1 - 3 210

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
exocytosis
positive regulation of cell proliferation
synaptic vesicle docking
vesicle-mediated transport
neuron projection development
synaptic vesicle fusion to presynaptic active zone membrane
positive regulation of cell adhesion
intracellular transport
vesicle docking
positive regulation of chemotaxis
long-term synaptic potentiation
membrane fusion
positive regulation of protein localization to plasma membrane
positive regulation of protein localization to cell surface
SNARE binding
SNAP receptor activity
protein binding
arachidonic acid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0