This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARL6IP5 SIRNA Assays ›
Gene Symbol:
Arl6ip5
Gene Name:
ADP-ribosylation factor-like 6 interacting protein 5
Gene Aliases:
Gtrap3-18
Chromosome Location:
Chr.4: 129574363 - 129598240 on Build Rnor_6.0
UniGene ID:
Rn.64512
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 192340

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arl6ip5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023972.3 NP_076462.1 2 293
GenBank mRNA AF240182.1 - 2 293
BC061548.1 - 305 6922366
FQ213351.1 - 326
FQ213919.1 - 315
FQ216190.1 - 326
FQ216551.1 - 327
FQ225681.1 - 326
FQ228780.1 - 391
FQ232358.1 - 326

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intrinsic apoptotic signaling pathway in response to oxidative stress
negative regulation of mitochondrial membrane potential
L-glutamate transport
positive regulation of stress-activated MAPK cascade
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of transport
protein C-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0