This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PWP1 SIRNA Assays ›
Gene Symbol:
PWP1
Gene Name:
PWP1 homolog, endonuclein
Gene Aliases:
IEF-SSP-9502
Chromosome Location:
Chr.12: 107685732 - 107713162 on Build GRCh38
UniGene ID:
Hs.506652
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 19232

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PWP1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317962.1 NP_001304891.1 5, 6 527
NM_001317963.1 NP_001304892.1 5, 6 699
NM_007062.2 NP_008993.1 5, 6 671
GenBank mRNA AK223162.1 - 5, 6 595
AK290681.1 - 5, 6 512
AK296252.1 - 5, 6 528
AK297961.1 - 4, 5 356
BC000067.2 - 5, 6 554
BC010921.1 - 5, 6 633
BC032127.2 - 5, 6 593
BC040135.1 - 5, 6 569
BC046144.1 - 5, 6 585
L07758.1 - 5, 6 590
Z36805.1 - 2, 3 172

Back To Top

More Information


Gene Ontology Categories:

Function(s)

transcription, DNA-templated

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0