This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HNRNPF SIRNA Assays ›
Gene Symbol:
Hnrnpf
Gene Name:
heterogeneous nuclear ribonucleoprotein F
Gene Aliases:
Hnrpf
Chromosome Location:
Chr.4: 149957206 - 149978896 on Build Rnor_6.0
UniGene ID:
Rn.144596
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 192103

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hnrnpf
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037285.1 NP_001032362.1 4 1644
NM_001037286.1 NP_001032363.1 3 1686
NM_001037287.1 NP_001032364.1 4 1601
NM_022397.3 NP_071792.1 4 1615
XM_008763262.1 XP_008761484.1 1 1373
GenBank mRNA AB022209.1 - 1 1338
BC097275.1 - 4 1584 7444983
BC103634.1 - 4 1603 7097633

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133834, NM_001166427, NM_001166432, NM_001166430, NM_001166428, and others.

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
regulation of RNA splicing
nucleotide binding
single-stranded RNA binding
TBP-class protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0