This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CARD9 SIRNA Assays ›
Gene Symbol:
Card9
Gene Name:
caspase recruitment domain family, member 9
Gene Aliases:
-
Chromosome Location:
Chr.3: 3798346 - 3807911 on Build Rnor_6.0
UniGene ID:
Rn.64486
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 192087

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Card9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022303.1 NP_071639.1 2 205
XM_008761625.1 XP_008759847.1 2 1393
XM_008761626.1 XP_008759848.1 2 1393
GenBank mRNA AF311288.1 - 2 205

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

I-kappaB kinase/NF-kappaB signaling
response to fungus
response to peptidoglycan
response to muramyl dipeptide
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
positive regulation of stress-activated MAPK cascade
response to drug
regulation of tumor necrosis factor biosynthetic process
regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to exogenous dsRNA
regulation of interleukin-2 biosynthetic process
innate immune response
positive regulation of innate immune response
regulation of interleukin-6 biosynthetic process
positive regulation of JNK cascade
defense response to Gram-positive bacterium
positive regulation of NF-kappaB transcription factor activity
defense response to virus
protein domain specific binding
protein homodimerization activity
CARD domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0