This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EXOC5 SIRNA Assays ›
Gene Symbol:
Exoc5
Gene Name:
exocyst complex component 5
Gene Aliases:
Sec10l1
Chromosome Location:
Chr.15: 25888757 - 25933302 on Build Rnor_6.0
UniGene ID:
Rn.44430
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 192011

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Exoc5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022204.3 NP_071540.1 2, 3 342
GenBank mRNA BC128739.1 - 2, 3 345 8362342
FQ217124.1 - 2, 3 371
U79417.1 - 2, 3 114

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_207214, AK170148, AK164067, AK167145, AK157473, and others.

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
protein transport
vesicle docking
Ral GTPase binding
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0