This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IFIT1 SIRNA Assays ›
Gene Symbol:
Ifit1 Ifit1lb
Gene Name:
interferon-induced protein with tetratricopeptide repeats 1
interferon-induced protein with tetratricopeptide repeats 1-like B
Gene Aliases:
Garg16
Ifit1
Chromosome Location:
Chr.1: 252944105 - 252946170 on Build Rnor_6.0
Chromosome Location:
Chr.1: 252914684 - 252926426 on Build Rnor_6.0
UniGene ID:
Rn.48677
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 191875

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ifit1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020096.1 NP_064481.1 1 320
GenBank mRNA AJ276893.1 - 1 320
Gene Symbol: Ifit1lb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760355.1 XP_008758577.1 2 1785
XM_220058.5 XP_220058.2 2 2681

Back To Top

More Information


Gene Ontology Categories:

Function(s)

response to virus
cellular response to interferon-alpha
cellular response to interferon-beta

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header