This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PADI4 SIRNA Assays ›
Gene Symbol:
Padi4
Gene Name:
peptidyl arginine deiminase, type IV
Gene Aliases:
Pdi4
Chromosome Location:
Chr.5: 159260569 - 159293694 on Build Rnor_6.0
UniGene ID:
Rn.11155
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 191760

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Padi4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017227.1 NP_058923.1 2 268
XM_006239145.2 XP_006239207.1 2 289
GenBank mRNA AB008803.1 - 2 268
AB010999.1 - 2 268

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
protein citrullination
stem cell population maintenance
histone H3-R26 citrullination
histone citrullination
innate immune response
protein-arginine deiminase activity
calcium ion binding
arginine deiminase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0