This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NSMAF SIRNA Assays ›
Gene Symbol:
Nsmaf
Gene Name:
neutral sphingomyelinase (N-SMase) activation associated factor
Gene Aliases:
FAN
Chromosome Location:
Chr.5: 19499111 - 19560304 on Build Rnor_6.0
UniGene ID:
Rn.229389
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 191506

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nsmaf
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181389.2 NP_852054.1 5, 6 349
XM_008763544.1 XP_008761766.1 5, 6 1408
XM_008763545.1 XP_008761767.1 5, 6 1408
GenBank mRNA AY163240.1 - 5, 6 349

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

endosomal transport
positive regulation of apoptotic process
positive regulation of catalytic activity
phospholipid binding
sphingomyelin phosphodiesterase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0