This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BPIFA1 SIRNA Assays ›
Gene Symbol:
Bpifa1
Gene Name:
BPI fold containing family A, member 1
Gene Aliases:
Plunc
Chromosome Location:
Chr.3: 149624867 - 149630610 on Build Rnor_6.0
UniGene ID:
Rn.19272
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 191344

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bpifa1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172031.1 NP_742028.1 2 221
GenBank mRNA AF393750.1 - 2 221

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011126, U69172, BC054375, BY121014.

Gene Ontology Categories:

Function(s) Process(es)

antibacterial humoral response
innate immune response
regulation of liquid surface tension
multicellular organismal water homeostasis
negative regulation of single-species biofilm formation in or on host organism
regulation of sodium ion transmembrane transport
molecular_function
lipid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header