This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WBP2 SIRNA Assays ›
Gene Symbol:
Wbp2
Gene Name:
WW domain binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.10: 104629568 - 104637903 on Build Rnor_6.0
UniGene ID:
Rn.198920
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 191119

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wbp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138975.1 NP_620431.1 2 154
XM_006247705.2 XP_006247767.1 2 234
GenBank mRNA AF499026.1 - 2 154

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

establishment of protein localization
positive regulation of gene expression, epigenetic
positive regulation of transcription from RNA polymerase II promoter
cellular response to estrogen stimulus
positive regulation of histone H3-K14 acetylation
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II transcription coactivator activity
protein binding
chromatin DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0