This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNJ15 SIRNA Assays ›
Gene Symbol:
Kcnj15
Gene Name:
potassium channel, inwardly rectifying subfamily J, member 15
Gene Aliases:
Kir4.2
Chromosome Location:
Chr.11: 35545188 - 35588517 on Build Rnor_6.0
UniGene ID:
Rn.81018
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 190849

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnj15
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133321.2 NP_579855.1 3 675
XM_006248128.2 XP_006248190.1 4 764
XM_006248129.2 XP_006248191.1 4 901
XM_006248130.2 XP_006248192.1 5 1063
XM_006248131.2 XP_006248193.1 4 904
XM_006248132.2 XP_006248194.1 4 855
XM_006248133.2 XP_006248195.1 3 639
XM_006248135.2 XP_006248197.1 3 1096
XM_006248137.2 XP_006248199.1 2 917
XM_006248138.2 XP_006248200.1 2 556
XM_006248139.2 XP_006248201.1 4 912
XM_006248141.2 XP_006248203.1 2 1786
XM_008768550.1 XP_008766772.1 4 872
XM_008768551.1 XP_008766773.1 4 826
XM_008768552.1 XP_008766774.1 4 783
XM_008768553.1 XP_008766775.1 3 3039
GenBank mRNA AY028455.1 - 3 676

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
potassium ion import
regulation of ion transmembrane transport
potassium ion transmembrane transport
inward rectifier potassium channel activity
potassium channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0