This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHEK1 SIRNA Assays ›
Gene Symbol:
Chek1
Gene Name:
checkpoint kinase 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 39181162 - 39202647 on Build Rnor_6.0
UniGene ID:
Rn.33267
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 190673

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chek1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080400.1 NP_536325.1 4 379
XM_008766042.1 XP_008764264.1 4 1512
XM_008766043.1 XP_008764265.1 4 449
GenBank mRNA AF414135.1 - 4 379
AF443592.1 - 4 377

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA damage checkpoint
G2/M transition of mitotic cell cycle
DNA repair
protein phosphorylation
cellular response to DNA damage stimulus
DNA damage induced protein phosphorylation
regulation of gene expression
regulation of double-strand break repair via homologous recombination
regulation of transcription from RNA polymerase II promoter in response to UV-induced DNA damage
negative regulation of G2/M transition of mitotic cell cycle
peptidyl-threonine phosphorylation
histone H3-T11 phosphorylation
regulation of cell proliferation
negative regulation of mitotic nuclear division
regulation of mitotic centrosome separation
chromatin-mediated maintenance of transcription
cellular response to mechanical stimulus
cellular response to organic substance
cellular response to caffeine
negative regulation of DNA biosynthetic process
regulation of histone H3-K9 acetylation
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
histone kinase activity (H3-T11 specific)

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0