This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SEC14L2 SIRNA Assays ›
Gene Symbol:
Sec14l2
Gene Name:
SEC14-like 2 (S. cerevisiae)
Gene Aliases:
Spf
Chromosome Location:
Chr.14: 84335594 - 84360135 on Build Rnor_6.0
UniGene ID:
Rn.46850
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 190454

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sec14l2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053801.2 NP_446253.2 9 734
XM_006251154.2 XP_006251216.1 9 723
GenBank mRNA AF309558.1 - 9 691
BC089785.1 - 9 734 7322689

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transport
positive regulation of catalytic activity
positive regulation of cholesterol biosynthetic process
transporter activity
enzyme activator activity
lipid binding
transferase activity, transferring alkyl or aryl (other than methyl) groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0