This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DVL1 SIRNA Assays ›
Gene Symbol:
Dvl1
Gene Name:
dishevelled segment polarity protein 1
Gene Aliases:
dvl-1
Chromosome Location:
Chr.5: 173295955 - 173308014 on Build Rnor_6.0
UniGene ID:
Rn.144610
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 190018

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dvl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031820.1 NP_114008.1 5 578
XM_006239598.2 XP_006239660.1 5 892
XM_008764388.1 XP_008762610.1 5 893
XM_008764389.1 XP_008762611.1 5 893
XM_008764390.1 XP_008762612.1 5 893
XM_008764391.1 XP_008762613.1 6 594
GenBank mRNA AF143545.2 - 5 578
AF143546.2 - 5 578
BC062070.1 - 4 604

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010091, BC138849, BC138848, AK155349, AK220168, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of neurotransmitter levels
regulation of protein phosphorylation
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
transcription from RNA polymerase II promoter
negative regulation of protein kinase activity
neurotransmitter secretion
axonogenesis
axon guidance
neuromuscular junction development
positive regulation of neuron projection development
Wnt signaling pathway
neural tube development
convergent extension involved in neural plate elongation
positive regulation of Wnt signaling pathway
cytoplasmic microtubule organization
negative regulation of protein binding
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein localization to nucleus
social behavior
protein localization to microtubule
intracellular signal transduction
receptor clustering
positive regulation of transcription, DNA-templated
collateral sprouting
axon extension
dendrite morphogenesis
synapse organization
convergent extension involved in organogenesis
canonical Wnt signaling pathway
Wnt signaling pathway, planar cell polarity pathway
prepulse inhibition
skeletal muscle acetylcholine-gated channel clustering
cochlea morphogenesis
planar cell polarity pathway involved in neural tube closure
positive regulation of canonical Wnt signaling pathway
frizzled binding
enzyme binding
protein kinase binding
identical protein binding
Rac GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0