This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HSD17B4 SIRNA Assays ›
Gene Symbol:
Hsd17b4
Gene Name:
hydroxysteroid (17-beta) dehydrogenase 4
Gene Aliases:
MPF-2
Chromosome Location:
Chr.18: 44810462 - 44897677 on Build Rnor_6.0
UniGene ID:
Rn.2082
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 189621

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hsd17b4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024392.2 NP_077368.2 7, 8 477
GenBank mRNA BC072472.1 - 7, 8 467 7100430
S83279.1 - 7, 8 449
U37486.1 - 7, 8 477
X94978.1 - 7, 8 452

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
osteoblast differentiation
fatty acid beta-oxidation
metabolic process
cholesterol metabolic process
androgen metabolic process
estrogen metabolic process
very long-chain fatty-acyl-CoA metabolic process
medium-chain fatty-acyl-CoA metabolic process
oxidation-reduction process
Sertoli cell development
3-hydroxyacyl-CoA dehydrogenase activity
receptor binding
long-chain-enoyl-CoA hydratase activity
isomerase activity
testosterone dehydrogenase [NAD(P)] activity
3alpha,7alpha,12alpha-trihydroxy-5beta-cholest-24-enoyl-CoA hydratase activity
protein homodimerization activity
17-beta-hydroxysteroid dehydrogenase (NAD+) activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header