This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC13A3 SIRNA Assays ›
Gene Symbol:
Slc13a3
Gene Name:
solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3
Gene Aliases:
Nadc3
Chromosome Location:
Chr.3: 162084336 - 162147398 on Build Rnor_6.0
UniGene ID:
Rn.3587
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 189487

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc13a3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022866.2 NP_074057.2 13 2550
GenBank mRNA AF080451.1 - 13 2526
AF081825.1 - 13 2553
BC129094.1 - 13 2550 8365494

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

sodium ion transport
dicarboxylic acid transport
succinate transport
citrate transport
aspartate transport
sodium ion transmembrane transport
transmembrane transport
succinate transmembrane transport
L-aspartate transmembrane transport
dicarboxylic acid transmembrane transporter activity
organic acid:sodium symporter activity
citrate transmembrane transporter activity
succinate transmembrane transporter activity
L-aspartate transmembrane transporter activity
high-affinity sodium:dicarboxylate symporter activity
sodium:dicarboxylate symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0