This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPIB SIRNA Assays ›
Gene Symbol:
Ppib
Gene Name:
peptidylprolyl isomerase B
Gene Aliases:
CypB, Scylp
Chromosome Location:
Chr.8: 71719681 - 71725538 on Build Rnor_6.0
UniGene ID:
Rn.1893
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 189405

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppib
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022536.2 NP_071981.1 5 591
GenBank mRNA AF071225.1 - 5 591
BC061971.1 - 5 620 5623342
FQ211358.1 - 5 595
FQ212031.1 - 5 620
FQ219792.1 - 5 629
FQ219926.1 - 5 641
FQ221020.1 - 5 644
FQ221248.1 - 5 639
FQ221362.1 - 5 618
FQ222696.1 - 5 753
FQ223162.1 - 5 638
FQ228087.1 - 5 634
FQ229174.1 - 5 641
FQ229214.1 - 5 635
FQ230004.1 - 645
FQ231656.1 - 5 634
FQ232077.1 - 5 630
FQ232920.1 - 5 644

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
positive regulation of multicellular organism growth
positive regulation by host of viral process
positive regulation by host of viral genome replication
protein stabilization
nuclear transport
bone development
chaperone-mediated protein folding
regulation of post-translational protein modification
peptidyl-prolyl cis-trans isomerase activity
protein binding
protein complex binding
peptide binding
poly(A) RNA binding
RNA polymerase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0