This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAD50 SIRNA Assays ›
Gene Symbol:
Rad50
Gene Name:
RAD50 homolog (S. cerevisiae)
Gene Aliases:
-
Chromosome Location:
Chr.10: 39002130 - 39054042 on Build Rnor_6.0
UniGene ID:
Rn.51136
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 189313

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rad50
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022246.1 NP_071582.1 16, 17 2881
GenBank mRNA AF218576.1 - 16, 17 2881
FQ232550.1 - 14, 15 2405

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of mitotic recombination
telomere maintenance
DNA catabolic process, endonucleolytic
DNA repair
double-strand break repair
DNA recombination
cellular response to DNA damage stimulus
telomere maintenance via telomerase
reciprocal meiotic recombination
regulation of mitotic cell cycle
positive regulation of protein autophosphorylation
DNA duplex unwinding
positive regulation of kinase activity
negative regulation of viral entry into host cell
chromosome organization
protein heterooligomerization
chromosome organization involved in meiotic cell cycle
nucleic acid phosphodiester bond hydrolysis
single-stranded DNA endodeoxyribonuclease activity
DNA binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
3'-5' exonuclease activity
ATPase activity
protein binding, bridging
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0