This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other XLR5C SIRNA Assays ›
Gene Symbol:
Xlr5c Xlr5a Xlr5b
Gene Name:
X-linked lymphocyte-regulated 5C
X-linked lymphocyte-regulated 5A
X-linked lymphocyte-regulated 5B
Gene Aliases:
Xlr5
Xlr5d
OTTMUSG00000017646
Chromosome Location:
Chr.X: 73283361 - 73293970 on Build GRCm38
Chromosome Location:
Chr.X: 73107621 - 73119524 on Build GRCm38
Chromosome Location:
Chr.X: 73146976 - 73158886 on Build GRCm38
UniGene ID:
Mm.255790
Mm.435653
Mm.255790
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
siRNA ID 188940

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Xlr5c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031493.1 NP_113681.1 6 475
GenBank mRNA BC119509.2 - 6 475 40053468
Gene Symbol: Xlr5a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045539.2 NP_001039004.2 7 607
GenBank mRNA BC051681.1 - 7 607 30021721
Gene Symbol: Xlr5b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111293.1 NP_001104763.1 7 562

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

spermatid development
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0